2,6-Difluoro-3-iodopyridine 95%
Supplier: Ambeed
|
Warning
|
A166290-100G
A166290-10G
A166290-1G
A166290-25G
A166290-5G
103806-044EA
18
USD
103806-044
103844-304
76813-020
76813-022
77320-550
77320-552
77320-554
2,6-Difluoro-3-iodopyridine 95%
2,6-Difluoro-3-iodopyridine
Precaución: for Research Use Only
Formula:
C₅H₂F₂IN MW: 240.98 g/mol Boiling Pt: 230.1 °C (760 mm Hg) Flash Pt: 92.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09743595 CAS Number: 685517-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...