2,6-Difluoro-3-(propane-1-sulfonamido)benzoic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,6-difluoro-3-(propane-1-sulphonamido)benzoic acid, 2,6-Difluoro-3-(propylsulphonamido)benzoic acid
A154791-100G
A154791-250MG
A154791-1G
A154791-5G
103804-678EA
13
USD
103804-678
103842-870
76871-794
76871-796
77318-612
77318-614
2,6-Difluoro-3-(propane-1-sulfonamido)benzoic acid 97%
2,6-Difluoro-3-(propane-1-sulfonamido)benzoic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₁F₂NO₄S MW: 279.26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18157658 CAS Number: 1103234-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...