2,6-Diethylaniline 98%
Supplier: Ambeed
|
Warning
|
A119195-100G
A119195-25G
A119195-500G
A119195-1000G
103808-200EA
39
USD
103808-200
77246-024
77246-026
77246-028
77297-664
2,6-Diethylaniline 98%
2,6-Diethylaniline
Precaución: for Research Use Only
Formula:
C₁₀H₁₅N MW: 149.24 g/mol Boiling Pt: 243 °C (760 mm Hg) Melting Pt: 3…4 °C Flash Pt: 115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007753 CAS Number: 579-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...