2,6-Dichloropyridine-3,4-diamine 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Diamino-2,6-dichloropyridine

A197570-100MG A197570-10G A197570-25G A197570-5G
103816-942EA 15 USD
103816-942 103847-562 76785-170 76785-172 77085-574 77085-576
2,6-Dichloropyridine-3,4-diamine 98%
2,6-Dichloropyridine-3,4-diamine

Precaución: for Research Use Only

Formula: C₅H₅Cl₂N₃
MW: 178.0200 g/mol
Boiling Pt: 424 °C (760 mm Hg)
Melting Pt: 181…183 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10697689
CAS Number: 101079-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR