4-Amino-2,6-dichloropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dichloro-4-pyridylamine, 2,6-Dichloropyridin-4-amine

A105616-100G A105616-10G A105616-25G A105616-500G A105616-1G A105616-5G
103796-912EA 6 USD
103796-912 103822-648 76982-386 76982-388 76982-390 76982-392 77309-514 77309-516
4-Amino-2,6-dichloropyridine 98%
4-Amino-2,6-dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Boiling Pt: 336.7 °C (760 mm Hg)
Melting Pt: 169…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00052832
CAS Number: 2587-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR