2,6-Dichloropyridin-3-ol 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dichloro-3-hydroxypyridine

A147733-100MG A147733-250MG
103803-820EA 110 USD
103803-820 76984-518 76984-520
2,6-Dichloropyridin-3-ol 98%
2,6-Dichloropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃Cl₂NO
MW: 163.99 g/mol
Boiling Pt: 353.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11044319
CAS Number: 52764-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR