2,6-Dichlorophenylhydrazinium chloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,6-Dichlorophenylhydrazine hydrochloride, 1-(2,6-Dichlorophenyl)hydrazine hydrochloride
A333290-100G
A333290-500G
A333290-25G
A333290-5G
103836-594EA
9
USD
103836-594
77114-932
77114-934
77306-444
77306-446
2,6-Dichlorophenylhydrazinium chloride 98%
2,6-Dichlorophenylhydrazinium chloride
Precaución: for Research Use Only
Formula:
C₆H₇Cl₃N₂ MW: 213.49 g/mol Boiling Pt: 253.5 °C (760 mm Hg) Melting Pt: 225 °C Flash Pt: 107.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012930 CAS Number: 50709-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...