2,6-Dichlorodiphenylamine 98%
Supplier: Ambeed
|
Warning
|
A241841-100G
A241841-10G
A241841-500G
A241841-1000G
A241841-25G
A241841-5G
103809-712EA
150
USD
103809-712
103851-242
77249-792
77249-794
77249-796
77304-224
77304-226
77304-228
2,6-Dichlorodiphenylamine 98%
2,6-Dichlorodiphenylamine
Precaución: for Research Use Only
Formula:
C₁₂H₉Cl₂N Boiling Pt: 309.8 °C (760 mm Hg) Melting Pt: 50…53 °C Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00269648 CAS Number: 15307-93-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...