2,6-Dichloronicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dichloro-3-pyridinecarboxylic acid, 2,6-Dichloropyridine-3-carboxylic acid

A109676-100G A109676-10G A109676-1G A109676-25G A109676-500G A109676-5G
103823-346EA 9 USD
103823-346 77270-786 77270-788 77270-790 77270-792 77270-794 77310-152
2,6-Dichloronicotinic acid 98%
2,6-Dichloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Boiling Pt: 351.2 °C (760 mm Hg)
Melting Pt: 140…143 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075583
CAS Number: 38496-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR