2,6-Dichloronicotinamide 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dichloropyridine-3-carboxamide

A107546-250MG A107546-25G A107546-1G A107546-5G
103797-210EA 7 USD
103797-210 103822-978 76821-620 76821-622 77309-834 77309-836
2,6-Dichloronicotinamide 97%
2,6-Dichloronicotinamide

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O
MW: 191.015 g/mol
Boiling Pt: 315.5±42.0 °C (760 mm Hg)
Melting Pt: 148…151 °C
Flash Pt: 144.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03230010
CAS Number: 62068-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR