2,6-Dichloroisonicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dichloropyridine-4-carboxylic acid

A173281-100G A173281-10G A173281-500G A173281-1G A173281-25G A173281-5G
103806-810EA 12 USD
103806-810 103845-104 77151-382 77151-384 77151-386 77301-254 77301-256 77301-258
2,6-Dichloroisonicotinic acid 97%
2,6-Dichloroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Boiling Pt: 437.8 °C (760 mm Hg)
Melting Pt: 210…211 °C
Flash Pt: 218.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00234147
CAS Number: 5398-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR