2,6-Dichlorobenzoic acid 98%

Supplier: Ambeed
Warning

A156637-1000G A156637-100G A156637-10G A156637-500G A156637-5G A156637-25G
103843-110EA 5 USD
103843-110 77110-506 77110-508 77110-510 77110-512 77300-204 77318-906
2,6-Dichlorobenzoic acid 98%
2,6-Dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: 298.7 °C (760 mm Hg)
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00002418
CAS Number: 50-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR