2,6-Dichloro-9-methyl-9H-purine 97%

Supplier: Ambeed
Warning

A119132-10G A119132-250MG A119132-25G A119132-5G
103799-170EA 19 USD
103799-170 77026-270 77026-272 77026-274 77311-846
2,6-Dichloro-9-methyl-9H-purine 97%
2,6-Dichloro-9-methyl-9H-purine

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₄
MW: 203.02 g/mol
Melting Pt: 152…153 °C
Flash Pt: 139.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00278778
CAS Number: 2382-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR