2,6-Dichloro-5-fluoronicotinic acid 98%
Supplier: Ambeed
|
Warning
|
A123716-100G
A123716-10G
A123716-1G
A123716-25G
A123716-500G
A123716-5G
77284-982EA
41
USD
77284-982
77284-984
77284-986
77284-988
77284-990
77312-554
2,6-Dichloro-5-fluoronicotinic acid 98%
2,6-Dichloro-5-fluoronicotinic acid
Precaución: for Research Use Only
Formula:
C₆H₂Cl₂FNO₂ Melting Pt: 152…155 °C Flash Pt: 102.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00799517 CAS Number: 82671-06-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...