2,6-Dichloro-5-fluoropyridin-3-amine 98%

Supplier: Ambeed
Warning

A368823-1G A368823-25G A368823-5G A368823-100MG A368823-10G A368823-250MG
103826-442EA 15 USD
103826-442 103837-456 76840-700 76840-702 76840-704 76978-668 76978-670 76978-672
2,6-Dichloro-5-fluoropyridin-3-amine 98%
2,6-Dichloro-5-fluoropyridin-3-amine

Precaución: For research use only

Formula: C₅H₃Cl₂FN₂
MW: 180.995 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11044255
CAS Number: 152840-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR