2,6-Dichloro-4-nitropyridine 97%
Supplier: Ambeed
|
Danger
|
A152200-100G
A152200-10G
A152200-250MG
A152200-25G
A152200-5G
A152200-1G
103804-358EA
16
USD
103804-358
103842-518
77235-468
77235-470
77235-472
77299-932
77299-934
77318-132
2,6-Dichloro-4-nitropyridine 97%
2,6-Dichloro-4-nitropyridine
Precaución: for Research Use Only
Formula:
C₅H₂Cl₂N₂O₂ MW: 192.99 g/mol Boiling Pt: 282.9 °C (760 mm Hg) Melting Pt: 94…98 °C Flash Pt: 124.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD05670545 CAS Number: 25194-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...