2,6-Dichloro-4-methyl-3-nitropyridine 98%

Supplier: Ambeed
Warning

A455312-1G A455312-5G A455312-100MG A455312-10G A455312-250MG
103827-838EA 28 USD
103827-838 103838-908 76842-754 76842-756 77252-856 77252-858 77252-860
2,6-Dichloro-4-methyl-3-nitropyridine 98%
2,6-Dichloro-4-methyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O₂
Boiling Pt: 316.9 °C (760 mm Hg)
Flash Pt: 145 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11110694
CAS Number: 60010-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR