2,6-Dichloro-4-iodopyridine 98%

Supplier: Ambeed
Warning

A193134-25G A193134-5G A193134-100G A193134-10G A193134-1G
103816-504EA 9 USD
103816-504 103847-118 77085-020 77085-022 77236-956 77236-958 77302-326
2,6-Dichloro-4-iodopyridine 98%
2,6-Dichloro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂CI₂IN
MW: 273.89 g/mol
Boiling Pt: 291.6 °C (760 mm Hg)
Melting Pt: 161…165 °C
Flash Pt: 130.145 °C
Storage Temperature: Ambient
MDL Number: MFCD07368400
CAS Number: 98027-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR