2,6-Dichloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidine 97%
Supplier: Ambeed
|
Warning
|
A104936-250MG
A104936-1G
A104936-5G
103796-802EA
13
USD
103796-802
103822-528
76821-506
77283-308
77283-310
2,6-Dichloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidine 97%
2,6-Dichloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₁₆H₂₀Cl₂N₆ MW: 367.28 g/mol Melting Pt: 249…251 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07779463 CAS Number: 7139-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...