2,6-Dichloro-4-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A436580-1G A436580-25G A436580-5G A436580-10G
103827-614EA 6 USD
103827-614 103838-678 76842-416 76842-418 76842-420 77252-538
2,6-Dichloro-4-(trifluoromethyl)pyridine 98%
2,6-Dichloro-4-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂Cl₂F₃N
Boiling Pt: 200.1 °C (760 mm Hg)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00042246
CAS Number: 39890-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR