2,6-Dichloro-4-(trifluoromethyl)nicotinonitrile 98%

Supplier: Ambeed
Warning

A256187-1G A256187-25G A256187-5G A256187-100G A256187-10G A256187-250MG
103812-246EA 7 USD
103812-246 103852-110 77091-594 77091-596 77091-598 77193-730 77193-732 77193-734
2,6-Dichloro-4-(trifluoromethyl)nicotinonitrile 98%
2,6-Dichloro-4-(trifluoromethyl)nicotinonitrile

Precaución: for Research Use Only

Formula: C₇HCl₂F₃N₂
Boiling Pt: 282.4 °C (760 mm Hg)
Melting Pt: 38 °C
Flash Pt: 124.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00084977
CAS Number: 13600-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR