2,6-Dichloro-4-(trifluoromethoxy)aniline 95%

Supplier: Ambeed
Warning

A149774-1G A149774-25G
103804-078EA 11 USD
103804-078 103842-206 77299-752 77317-716
2,6-Dichloro-4-(trifluoromethoxy)aniline 95%
2,6-Dichloro-4-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₄Cl₂F₃NO
MW: 246.02 g/mol
Melting Pt: 30…32 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00190128
CAS Number: 99479-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR