2,6-Dichloro-3-methylpyridine 98%

Supplier: Ambeed
Warning

A265152-10G A265152-250MG A265152-1G A265152-25G A265152-5G
103812-708EA 6 USD
103812-708 103852-602 76816-088 76816-090 77092-266 77092-268 77092-270
2,6-Dichloro-3-methylpyridine 98%
2,6-Dichloro-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂N
Flash Pt: 117.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00137847
CAS Number: 58584-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR