2,6-Dichloro-3H-pyrimidin-4-one ≥98%

Supplier: Ambeed
Warning

A531713-5G A531713-100MG A531713-250MG
103828-574EA 36 USD
103828-574 103839-686 77613-664 77673-418 77673-420
2,6-Dichloro-3H-pyrimidin-4-one ≥98%
2,6-Dichloro-3H-pyrimidin-4-one

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₄H₂Cl₂N₂O
MW: 164.9775 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00955562
CAS Number: 120977-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR