2,6-Dichloro-3-formylpyridine 98%
Supplier: Ambeed
|
Warning
|
A297891-5G
A297891-100G
A297891-10G
A297891-1G
A297891-25G
103814-160EA
6
USD
103814-160
103835-304
76837-992
77099-456
77099-458
77305-730
77305-732
2,6-Dichloro-3-formylpyridine 98%
2,6-Dichloro-3-formylpyridine
Precaución: for Research Use Only
Formula:
C₆H₃Cl₂NO Melting Pt: 74…75 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07369746 CAS Number: 55304-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...