2,6-Dichloro-3-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

A224953-100G A224953-1G A224953-25G A224953-5G A224953-10G
103819-146EA 14 USD
103819-146 103850-038 76975-956 77088-700 77088-702 77088-704 77612-186
2,6-Dichloro-3-(trifluoromethyl)pyridine 97%
2,6-Dichloro-3-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂Cl₂F₃N
Boiling Pt: 221 °C (760 mm Hg)
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00042245
CAS Number: 55304-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR