2,6-Dibromopyridine 97%

Supplier: Ambeed
Warning

A113878-100G A113878-10G A113878-25G A113878-500G A113878-1000G
103808-112EA 190 USD
103808-112 77271-092 77271-094 77271-096 77271-098 77310-966
2,6-Dibromopyridine 97%
2,6-Dibromopyridine

Precaución: for Research Use Only

Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 255.9 °C (760 mm Hg)
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00006223
CAS Number: 626-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR