2,6-Dibromopyridin-4-amine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2,6-dibromopyridine

A101388-250MG A101388-25G A101388-1G A101388-5G A101388-10G
103795-948EA 27 USD
103795-948 103821-632 77025-170 77025-172 77297-080 77297-082 77548-906
2,6-Dibromopyridin-4-amine 97%
2,6-Dibromopyridin-4-amine

Precaución: for Research Use Only

Formula: C₅H₄Br₂N₂
MW: 251.9100 g/mol
Boiling Pt: 373.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00234047
CAS Number: 39771-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR