3-Amino-2,6-dibromopyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dibromopyridin-3-amine

A214170-25G A214170-5G A214170-100G A214170-10G A214170-500G A214170-1G
103818-372EA 9 USD
103818-372 103849-080 77087-582 77087-584 77288-068 77288-070 77288-072 77303-326
3-Amino-2,6-dibromopyridine 97%
3-Amino-2,6-dibromopyridine

Precaución: for Research Use Only

Formula: C₅H₄Br₂N₂
MW: 251.9100 g/mol
Boiling Pt: 327.7 °C (760 mm Hg)
Melting Pt: 214…218 °C
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD00128862
CAS Number: 39856-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR