2,6-Dibromopyrazine 95%

Supplier: Ambeed
Warning

A104033-250MG A104033-1G A104033-5G A104033-10G A104033-25G A104033-100G
103796-644EA 6 USD
103796-644 103822-356 76780-666 77309-218 77309-220 77565-808 77565-810 77666-498
2,6-Dibromopyrazine 95%
2,6-Dibromopyrazine

Precaución: For research use only.

Formula: C₄H₂Br₂N₂
Boiling Pt: 233.6 °C (760 mm Hg)
Flash Pt: 95.1 °C
Storage Temperature: Freezer
MDL Number: MFCD08061601
CAS Number: 23229-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR