2,6-Dibromophenol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3-Dibromo-2-hydroxybenzene
A307460-1000G
A307460-100G
A307460-10G
A307460-25G
A307460-5G
A307460-500G
103835-682EA
10
USD
103835-682
76816-840
76816-842
76816-844
76838-428
76838-430
77672-552
2,6-Dibromophenol 98%
Precaución: for Research Use Only
Formula:
C₆H₄Br₂O MW: 251.91 g/mol Boiling Pt: 256.6 °C (760 mm Hg) Melting Pt: 53…56 °C Flash Pt: 255 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002152 CAS Number: 608-33-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...