2,6-Dibromonicotinaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dibromopyridine-3-carboxaldehyde

A117200-10G A117200-250MG A117200-1G A117200-5G
103798-844EA 55 USD
103798-844 103824-716 77245-952 77245-954 77311-502 77311-504
2,6-Dibromonicotinaldehyde 95%
2,6-Dibromonicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃Br₂NO
MW: 264.9 g/mol
Boiling Pt: 331.6 °C (760 mm Hg)
Flash Pt: 154.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10574710
CAS Number: 55304-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR