2,6-Dibromonaphthalene 98%

Supplier: Ambeed
Danger

A161432-10G A161432-250MG A161432-1G A161432-25G A161432-5G
103805-490EA 12 USD
103805-490 103843-728 77110-710 77110-712 77319-706 77319-708 77319-710
2,6-Dibromonaphthalene 98%
2,6-Dibromonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂
Boiling Pt: 339.1 °C (760 mm Hg)
Melting Pt: 163…166 °C
Flash Pt: 184.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01026462
CAS Number: 13720-06-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR