2,6-Dibromoisonicotinic acid 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,6-Dibromopyridine-4-carboxylic acid
A386279-1G
A386279-5G
A386279-10G
A386279-250MG
103826-796EA
13
USD
103826-796
103837-834
76841-188
76841-190
77003-692
77003-694
2,6-Dibromoisonicotinic acid 96%
2,6-Dibromoisonicotinic acid
Precaución: for Research Use Only
Formula:
C₆H₃Br₂NO₂ MW: 280.9000 g/mol Boiling Pt: 487.5 °C (760 mm Hg) Melting Pt: 183…187 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09743987 CAS Number: 2016-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...