2,6-Dibromodithieno[3,2-b:2',3'-d]thiophene 97%

Supplier: Ambeed
Warning

A260346-100MG A260346-250MG A260346-1G A260346-5G
103812-492EA 179 USD
103812-492 103852-372 76827-120 76827-122 77304-788 77304-790
2,6-Dibromodithieno[3,2-b:2',3'-d]thiophene 97%
2,6-Dibromodithieno[3,2-b:2',3'-d]thiophene

Precaución: for Research Use Only

Formula: C₈H₂Br₂S₃
Boiling Pt: 447.9 °C (760 mm Hg)
Melting Pt: 165…169 °C
Flash Pt: 224.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11215199
CAS Number: 67061-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR