2,6-Dibromobenzonitrile 95%

Supplier: Ambeed
Warning

A696418-10G A696418-250MG A696418-1G A696418-5G
103840-610EA 32 USD
103840-610 76791-050 76791-052 76844-828 76844-830
2,6-Dibromobenzonitrile 95%
2,6-Dibromobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃Br₂N
MW: 260.91 g/mol
Boiling Pt: 326.5 °C (760 mm Hg)
Flash Pt: 151.3 °C
Storage Temperature: Ambient
MDL Number: MFCD09834774
CAS Number: 6575-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR