2,6-Dibromobenzoic acid 98%

Supplier: Ambeed
Warning

A182783-10G A182783-250MG A182783-1G A182783-5G
103815-566EA 10 USD
103815-566 103846-124 77029-504 77029-506 77083-684 77083-686
2,6-Dibromobenzoic acid 98%
2,6-Dibromobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄Br₂O₂
Boiling Pt: 333.4 °C (760 mm Hg)
Melting Pt: 146…149 °C
Flash Pt: 155.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00234254
CAS Number: 601-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR