2,6-Dibromo-9,10-anthraquinone 96%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dibromoanthracene-9,10-dione

A357098-1G A357098-25G A357098-5G A357098-100G A357098-10G A357098-250MG
103826-184EA 6 USD
103826-184 103837-188 76840-336 76840-338 76840-340 77251-618 77251-620 77251-622
2,6-Dibromo-9,10-anthraquinone 96%
2,6-Dibromo-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₆Br₂O₂
MW: 366.0042 g/mol
Boiling Pt: 491.8 °C (760 mm Hg)
Melting Pt: 290 °C
Flash Pt: 172.19 °C
Storage Temperature: Ambient
MDL Number: MFCD08276344
CAS Number: 633-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR