2,6-Dibromoaniline 98%

Supplier: Ambeed
Warning

A383810-1G A383810-25G A383810-5G A383810-100G A383810-10G A383810-500G
103826-750EA 9 USD
103826-750 103837-786 76841-122 76841-124 76841-126 77115-540 77115-542 77115-544
2,6-Dibromoaniline 98%
2,6-Dibromoaniline

Precaución: for Research Use Only

Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Boiling Pt: 265.8 °C (760 mm Hg)
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00007638
CAS Number: 608-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR