2,6-Dibromo-4-nitropyridine 97%

Supplier: Ambeed
Warning

A248885-1G A248885-25G A248885-5G A248885-10G
103811-846EA 10 USD
103811-846 103851-680 77091-022 77091-024 77091-026 77098-630
2,6-Dibromo-4-nitropyridine 97%
2,6-Dibromo-4-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂Br₂N₂O₂
MW: 281.8896 g/mol
Flash Pt: 153.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07369947
CAS Number: 175422-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR