2,6-Dibromo-4-methylpyridine 98%

Supplier: Ambeed
Warning

A120920-10G A120920-25G A120920-1G A120920-5G A120920-250MG
103800-008EA 69 USD
103800-008 76781-746 76781-748 77312-116 77312-118 77666-734
2,6-Dibromo-4-methylpyridine 98%
2,6-Dibromo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅Br₂N
Boiling Pt: 282.8 °C (760 mm Hg)
Melting Pt: 77…79 °C
Flash Pt: 125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11036225
CAS Number: 73112-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR