2,6-Dibromo-4-methoxypyridine 96%

Supplier: Ambeed
Warning

A300821-5G A300821-100G A300821-10G A300821-250MG A300821-25G
103814-280EA 12 USD
103814-280 103835-430 76838-116 76977-724 76977-726 76977-728 77305-812
2,6-Dibromo-4-methoxypyridine 96%
2,6-Dibromo-4-methoxypyridine

Precaución: For research use only

Formula: C₆H₅Br₂NO
MW: 266.918 g/mol
Boiling Pt: 283 °C (760 mm Hg)
Melting Pt: 131…137 °C
Flash Pt: 124.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11036227
CAS Number: 117873-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR