2,6-Dibromo-4-tert-butylaniline 97%

Supplier: Ambeed
Warning

Synonyms: 4-(tert-Butyl)-2,6-dibromoaniline, 2,6-Dibromo-4-(tert-butyl)aniline

A335162-25G A335162-5G A335162-100G A335162-10G A335162-250MG A335162-500G A335162-1G
103825-684EA 7 USD
103825-684 103836-654 76839-664 76839-666 76989-326 76989-328 76989-330 76989-332 77306-482
2,6-Dibromo-4-tert-butylaniline 97%
2,6-Dibromo-4-tert-butylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₃Br₂N
MW: 307.03 g/mol
Boiling Pt: 288 °C (760 mm Hg)
Flash Pt: 128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06657952
CAS Number: 10546-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR