2,6-Dibromo-3-nitropyridine 97%

Supplier: Ambeed
Warning

A111854-100G A111854-10G A111854-25G A111854-500G A111854-5G
103823-734EA 29 USD
103823-734 76970-862 76970-864 76970-866 76970-868 77310-546
2,6-Dibromo-3-nitropyridine 97%
2,6-Dibromo-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂Br₂N₂O₂
MW: 281.8900 g/mol
Boiling Pt: 295.8 °C (760 mm Hg)
Melting Pt: 70…71 °C
Flash Pt: 132.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00233995
CAS Number: 55304-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR