2,6-Dibromo-3-methoxypyridine 97%

Supplier: Ambeed
Warning

A164095-100MG A164095-250MG A164095-1G
103805-814EA 25 USD
103805-814 103844-078 76985-182 76985-184 77320-218
2,6-Dibromo-3-methoxypyridine 97%
2,6-Dibromo-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅Br₂NO
MW: 266.918 g/mol
Melting Pt: 99…100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD26743441
CAS Number: 79491-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR