2,6-Dibromo-3-hydroxypyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dibromopyridin-3-ol

A202562-1G A202562-25G A202562-5G A202562-100G A202562-10G A202562-250MG
103817-400EA 27 USD
103817-400 103848-044 77086-154 77086-156 77086-158 77248-866 77248-868 77248-870
2,6-Dibromo-3-hydroxypyridine 95%
2,6-Dibromo-3-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₃Br₂NO
MW: 252.8900 g/mol
Boiling Pt: 369.3 °C (760 mm Hg)
Melting Pt: 158…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318479
CAS Number: 6602-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR