2,6-Diacetylpyridine 97%
Supplier: Ambeed
|
Warning
|
A106259-100G
A106259-10G
A106259-250MG
A106259-25G
A106259-500G
A106259-1G
A106259-5G
103797-018EA
8
USD
103797-018
103822-770
77148-492
77148-494
77148-496
77148-498
77148-500
77309-630
77309-632
2,6-Diacetylpyridine 97%
2,6-Diacetylpyridine
Precaución: for Research Use Only
Formula:
C₉H₉NO₂ Melting Pt: 79…82 °C Flash Pt: 131.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006304 CAS Number: 1129-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...