2,6-Bis(bromomethyl)naphthalene 97%

Supplier: Ambeed
Danger

A934054-5G A934054-100MG A934054-250MG A934054-1G
103841-414EA 286 USD
103841-414 76845-678 76993-936 76993-938 77308-524
2,6-Bis(bromomethyl)naphthalene 97%
2,6-Bis(bromomethyl)naphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₀Br₂
MW: 314.0200 g/mol
Boiling Pt: 386.3 °C (760 mm Hg)
Melting Pt: 183…187 °C
Flash Pt: 219.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00228693
CAS Number: 4542-77-2
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR