2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥97%, ee 99%

Supplier: Ambeed
Warning

A266987-10G A266987-250MG A266987-1G A266987-5G A266987-25G
103812-824EA 45 USD
103812-824 103852-726 77014-306 77014-308 77092-440 77304-966 77890-475
2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥97%, ee 99%
2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₂₃H₁₉N₃O₂
Boiling Pt: 600.4 °C (760 mm Hg)
Melting Pt: 171…175 °C
Flash Pt: 316.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01863586
CAS Number: 174500-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR