(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (-)-2,6-Bis[(4S)-4-(i-propyl)-2-oxazolin-2-yl]pyridine, (S,S)-2,2'-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline), 2,6-Bis((S)-4-isopropyl-4,5-dihydrooxazol-2-yl)pyridine, (S)-(i-Pr)-pybox

A159703-25G A159703-1G A159703-5G A159703-250MG
103805-276EA 22 USD
103805-276 103843-498 76973-486 77319-394 77319-396 77881-249
(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine ≥98%, ee 99%
(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₇H₂₃N₃O₂
Boiling Pt: 457.1 °C (760 mm Hg)
Melting Pt: 155…157 °C (760 mm Hg)
Flash Pt: 230.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191815
CAS Number: 118949-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR